![Glucose monohydrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/154/372/2a77eff0-786e-4449-9857-28d439f0c66e/640/2a77eff0-786e-4449-9857-28d439f0c66e.png)
Glucose monohydrate
![Glucose monohydrate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/154/372/2a77eff0-786e-4449-9857-28d439f0c66e/640/2a77eff0-786e-4449-9857-28d439f0c66e.png)
Synonym(s):
D-(+)-Glucose monohydrate, Dextrose monohydrate
Empirical Formula (Hill Notation):
C6H12O6 · H2O
CAS Number:
Molecular Weight:
198.17
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing