![Gluconasturtiin potassium salt phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/711/689/19b3ec38-de66-47e2-8570-f3b5591cd865/640/19b3ec38-de66-47e2-8570-f3b5591cd865.png)
Gluconasturtiin potassium salt
![Gluconasturtiin potassium salt phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/711/689/19b3ec38-de66-47e2-8570-f3b5591cd865/640/19b3ec38-de66-47e2-8570-f3b5591cd865.png)
Synonym(s):
2-Phenylethylglucosinolate potassium salt
Empirical Formula (Hill Notation):
C15H20KNO9S2
CAS Number:
Molecular Weight:
461.55
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing