![Glimepiride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/154/410/f31a1c86-eda2-4901-b405-c06b16142d96/640/f31a1c86-eda2-4901-b405-c06b16142d96.png)
Glimepiride
![Glimepiride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/154/410/f31a1c86-eda2-4901-b405-c06b16142d96/640/f31a1c86-eda2-4901-b405-c06b16142d96.png)
Synonym(s):
Glimepiride, trans-3-Ethyl-2,5-dihydro-4-methyl-N-[N-[4-[[[[(4-methylcyclohexyl)amino]carbonyl]amino]sulfonyl]phenyl]ethyl]-2-oxo-1H-pyrrole-1-carboxamide, HOE-490
Empirical Formula (Hill Notation):
C24H34N4O5S
CAS Number:
Molecular Weight:
490.62
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing