Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Giant-size molecular model components
Products
Brand
Sigma-Aldrich (2)
Color
Length (cm)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z134643
Giant-size molecular model components,
nitrogen centers, blue, trigonal-bipyramidal
Expand
View Pricing
Z134813
Giant-size molecular model components,
metal centers, silver, octahedral
Expand
View Pricing
Z134589
Giant-size molecular model components,
carbon centers, black, tetrahedral
Expand
View Pricing
Z134767
Giant-size molecular model components,
hydrogen centers, white, monovalent
Expand
View Pricing
Z136379
Giant-size molecular model components,
double bonds, red, 8cm/Å
Expand
View Pricing
Z136360
Giant-size molecular model components,
single bonds, black, 8cm/Å
Expand
View Pricing
Z134651
Giant-size molecular model components,
oxygen centers, red, tetrahedral
Expand
View Pricing
Z136387
Giant-size molecular model components,
C-H bonds, yellow, 8cm/Å
Expand
View Pricing