Skip to Content
Merck
γ-Cyclodextrin hydrate

γ-Cyclodextrin hydrate

γ-Cyclodextrin hydrate
Synonym(s):
γ-Schardinger dextrin, Cyclooctaamylose
Empirical Formula (Hill Notation):
C48H80O40 · xH2O
CAS Number:
Molecular Weight:
1297.12 (anhydrous basis)
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing