Ganciclovir
Synonym(s):
2-Amino-1,9-dihydro-9-[[2-hydroxy-1-(hydroxymethyl)ethoxy]methyl]-6H-purin-6-one, 9-(1,3-Dihydroxy-2-propoxymethyl)guanine, DHPG, Ganciclovir ready made solution, 2-Amino-1,9-dihydro-9-[[2-hydroxy-1-(hydroxymethyl)ethoxy]methyl]-6H-purin-6-one, Ganciclovir
Empirical Formula (Hill Notation):
C9H13N5O4
CAS Number:
Molecular Weight:
255.23
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Activity Spectrum
Agency
Format
Research Area
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing