Skip to Content
Merck
vibrant-m

Fosinopril impurity A

vibrant-m
Synonym(s):
(2S,4S)-4-Cyclohexyl-1-[[(R)-hydroxy(4-phenylbutyl)phosphoryl]acetyl]pyrrolidine-2-carboxylic acid
Empirical Formula (Hill Notation):
C23H34NO5P
Molecular Weight:
435.49

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade