Formoterol fumarate dihydrate
Synonym(s):
(R*,R*)-N-[2-Hydroxy-5-[1-hydroxy-2-[[2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]phenyl]formamide fumarate, Formoterol hemifumarate salt monohydrate
Empirical Formula (Hill Notation):
C19H24N2O4 · 0.5C4H4O4 · H2O
CAS Number:
Molecular Weight:
420.46
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2