
Formamidine disulfide dihydrochloride

Synonym(s):
Formamidine disulfide dihydrochloride
Empirical Formula (Hill Notation):
C2H6N4S2 · 2HCl
CAS Number:
Molecular Weight:
223.15
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing