
Fluticasone Related Compound A

Synonym(s):
6ALPHA,9-DIFLUORO-11BETA-HYDROXY- 16ALPHA-METHYL-3-OXO-17 (PROPANOYLOXY)ANDROSTA-1,4-DIEN- 17BETA-CARBO(THIOPEROXOIC)SO-ACID (Fluticasone Related Compound A) (Fluticasone Impurity B), ((6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-(propionyloxy)-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthrene-17-carbo(thioperoxoic)o-acid)
Empirical Formula (Hill Notation):
C24H30F2O6S
CAS Number:
Molecular Weight:
484.55
Products
Brand
Formula Weight
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing