Skip to Content
Merck
vibrant-m

Fluorometholone Related Compound A

vibrant-m
Synonym(s):
(6α,11β)-11,17-Dihydroxy-6-methylpregna-1,4-diene-3,20-dione, 11β,17α-Dihydroxy-6α-methylpregna-1,4-diene-3,20-dione
Empirical Formula (Hill Notation):
C22H30O4
CAS Number:
Molecular Weight:
358.47

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1