Skip to Content
Merck
Fludeoxyglucose Related Compound B United States Pharmacopeia (USP) Reference Standard

Fludeoxyglucose Related Compound B

Fludeoxyglucose Related Compound B United States Pharmacopeia (USP) Reference Standard
Synonym(s):
2-Chloro-2-deoxy-D-glucose, 2-CIDG
Empirical Formula (Hill Notation):
C6H11ClO5
CAS Number:
Molecular Weight:
198.60

Products

Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1