
Flucloxacillin impurity C

Synonym(s):
(+)-6-Aminopenicillanic acid, (2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-APA
Empirical Formula (Hill Notation):
C8H12N2O3S
CAS Number:
Molecular Weight:
216.26
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing