
Fentrazamide

Synonym(s):
4-(2-Chlorophenyl)-N-cyclohexyl-N-ethyl-4,5-dihydro-5-oxo-1H-tetrazole-1-carboxamide
Empirical Formula (Hill Notation):
C16H20ClN5O2
CAS Number:
Molecular Weight:
349.82
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing