Skip to Content
Merck
Fenhexamid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Fenhexamid

Fenhexamid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
N-(2,3-Dichloro-4-hydroxyphenyl)-1-methylcyclohexanecarboxamide
Empirical Formula (Hill Notation):
C14H17Cl2NO2
CAS Number:
Molecular Weight:
302.20

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique