
Fast Red Violet LB Salt

Synonym(s):
5-Chloro-4-benzamido-2-methylbenzenediazonium chloride hemi(zinc chloride) salt
Linear Formula:
C14H11Cl2N3O · ½ZnCl2
CAS Number:
Molecular Weight:
376.31
Products
Product Category
Brand
Formula Weight
Physical Form
Chemical Composition
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing