Exo-2-Naphthyl Kwon [2.2.1] Bicyclic Phosphine
Synonym(s):
(1S,4S,5R)-5-(naphthalen-2-yl)-2-tosyl-2-aza-5-phosphabicyclo[2.2.1]heptane
Empirical Formula (Hill Notation):
C22H22NO2PS
Molecular Weight:
395.45
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing