Skip to Content
Merck
Ethyl sulfate sodium salt 1.0 mg/mL in methanol (as ethyl sulfate), ampule of 1 mL, certified reference material, Cerilliant®

Ethyl sulfate sodium salt

Ethyl sulfate sodium salt 1.0 mg/mL in methanol (as ethyl sulfate), ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C2H5O4SNa
CAS Number:
Molecular Weight:
148.11
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing