Skip to Content
Merck
Ethyl salicylate ≥99%, FCC, FG

Ethyl salicylate

Ethyl salicylate ≥99%, FCC, FG
Synonym(s):
Ethyl 2-hydroxybenzoate
Linear Formula:
2-(HO)C6H4CO2C2H5
CAS Number:
Molecular Weight:
166.17
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing