Skip to Content
Merck
Ethyl nonanoate ≥98%, FCC, FG

Ethyl nonanoate

Ethyl nonanoate ≥98%, FCC, FG
Synonym(s):
Ethyl pelargonate, Ethyl nonanoate
Linear Formula:
CH3(CH2)7COOC2H5
CAS Number:
Molecular Weight:
186.29

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing