Skip to Content
Merck
Ethylenediaminetetraacetic acid calcium disodium salt hydrate ≥97.0% (calc. based on dry substance, KT)

Ethylenediaminetetraacetic acid calcium disodium salt hydrate

Ethylenediaminetetraacetic acid calcium disodium salt hydrate ≥97.0% (calc. based on dry substance, KT)
Synonym(s):
Calcium disodium ethylenediaminetetraacetate hydrate
Linear Formula:
Ca(OOCCH2)2NCH2CH2N(CH2COONa)2 · aq
CAS Number:
Molecular Weight:
374.27 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reagent Type