
Ethylenediamine solution

Synonym(s):
1,2-Diaminoethane
Linear Formula:
NH2CH2CH2NH2
CAS Number:
Molecular Weight:
60.10
Products
Product Category
Formula Weight
Gene Alias
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing