Ethyl 3-ethoxyacrylate
Synonym(s):
Ethyl 3-ethoxy-2-propenoate
Linear Formula:
CH3CH2OCH=CHCOOCH2CH3
CAS Number:
Molecular Weight:
144.17
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing