Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Mastercycler
®
nexus
Products
Product Category
thermocyclers (8)
heaters (7)
Feature
Manufacturer
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
EP6331000017
Eppendorf
®
Mastercycler
®
nexus,
Gradient, complete unit, aluminum block, AC/DC input 230 V, 50—60 Hz, EuroPlug
Expand
View Pricing
EP6332000010
Eppendorf
®
Mastercycler
®
nexus,
Eco, satellite unit, aluminum block, AC/DC input 230 V (EU plug)
Expand
View Pricing
EP6334000018
Eppendorf
®
Mastercycler
®
nexus,
Gradient Eco, satellite unit, aluminum block, AC/DC input 230 V (EU plug)
Expand
View Pricing
EP6331000041
Eppendorf
®
Mastercycler
®
nexus,
Gradient, complete unit, aluminum block, AC/DC input 230V (GB plug)
Expand
View Pricing
EP6333000049
Eppendorf
®
Mastercycler
®
nexus,
Nexus, complete unit, aluminum block, AC/DC input 230V (GB plug)
Expand
View Pricing
EP6332000045
Eppendorf
®
Mastercycler
®
nexus,
Eco, satellite unit, aluminum block, AC/DC input 230V (GB)
Expand
View Pricing
EP6334000042
Eppendorf
®
Mastercycler
®
nexus,
Gradient Eco, satellite unit, aluminum block, AC/DC input 230V (GB plug)
Expand
View Pricing
EP6347000041
Eppendorf
®
Mastercycler
®
nexus,
GSX1e, complete unit, aluminum block, AC/DC input 230V (GB plug)
Expand
View Pricing