Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Deepwell plates, standard, 384 wells
Products
Product Category
plates (4)
Manufacturer
Wells
Color
Package Size
Sterility
Sterilization Method
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
EP0030521102
Eppendorf
®
Deepwell plates, standard, 384 wells,
white plate, conical bottom colorless wells, capacity 200 μL, pkg of 40 ea
Expand
View Pricing
EP0030525108
Eppendorf
®
Deepwell plates, standard, 384 wells,
white plate, conical bottom colorless wells, capacity 200 μL, pkg of 120 ea
Expand
View Pricing
EP0030522109
Eppendorf
®
Deepwell plates, standard, 384 wells,
white plate, conical bottom colorless wells, capacity 200 μL, sterile, pkg of 40 ea
Expand
View Pricing
EP0030526104
Eppendorf
®
Deepwell plates, standard, 384 wells,
white plate, conical bottom colorless wells, capacity 200 μL, sterile, pkg of 120 ea
Expand
View Pricing