Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Conical Tubes with SafeCode system (data matrix code and plain text labels)
Products
Product Category
tubes (3)
Manufacturer
Capacity (mL)
Color
Package Size
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
EP0030113500
Eppendorf
®
Conical Tubes with SafeCode system (data matrix code and plain text labels),
sterile, screw cap, capacity 5.0 mL, PCR clean, pack of 2 × 100 ea
Expand
View Pricing
EP0030113497
Eppendorf
®
Conical Tubes with SafeCode system (data matrix code and plain text labels),
non-sterile, screw cap, capacity 50 mL, PCR clean, pack of 20 × 25 ea
Expand
View Pricing
EP0030113489
Eppendorf
®
Conical Tubes with SafeCode system (data matrix code and plain text labels),
non-sterile, screw cap, capacity 15 mL, PCR clean, pack of 10 × 50 ea
Expand
View Pricing