Skip to Content
Merck
Epicatechin gallate primary reference standard

(−)-Epicatechin gallate

Epicatechin gallate primary reference standard
Synonym(s):
(−)-cis-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol 3-gallate, (−)-cis-3,3′,4′,5,7-Pentahydroxyflavane 3-gallate, 3-O-Galloylepicatechin, (−)-Epicatechin gallate
Empirical Formula (Hill Notation):
C22H18O10
CAS Number:
Molecular Weight:
442.37

Products

Product Category
Brand
Biological Source
Feature
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Research Area
Application
Reference Material Type
Special Grade