Skip to Content
Merck
EPA 8270 Base/Neutrals Surrogate Spike Mix HC certified reference material, 5000 μg/mL each component in dichloromethane

EPA 8270 Base/Neutrals Surrogate Spike Mix HC

EPA 8270 Base/Neutrals Surrogate Spike Mix HC certified reference material, 5000 μg/mL each component in dichloromethane
Synonym(s):
EPA 8270 Base/Neutrals Surrogate Spike Mix

Products

Brand
Product Line
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique