Skip to Content
AT
EN
Back
Ectoine
Synonym(s):
(
S
)-2-Methyl-1,4,5,6-tetrahydropyrimidine-4-carboxylic acid, Thp(B)
Empirical Formula (Hill Notation):
C
6
H
10
N
2
O
2
CAS Number:
96702-03-3
Molecular Weight:
142.16
EC Number:
431-910-1
Products
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
81619
Ectoine,
≥95.0% (HPLC)
Expand
View Pricing
E2271
Ectoine,
osmoprotectant
Expand
View Pricing
02380595
Ectoine,
primary reference standard
Expand
View Pricing