Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Durapore
®
Screen Filter
Products
Product Category
filters (4)
membranes (4)
Brand
Millipore (4)
Product Line
Diameter (mm)
Hydrophobicity
Color
Membrane Material
Pore size (µm)
Sterility
Sterilization Method
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
GVWP2932A
Durapore
®
Screen Filter,
pore size 0.22 μm, PVDF membrane, hydrophilic, filter diam. 293 mm
Expand
View Pricing
HVLP2932A
Durapore
®
Screen Filter,
pore size 0.45 μm, PVDF membrane, hydrophilic, filter diam. 293 mm
Expand
View Pricing
GVHP29325
Durapore
®
Screen Filter,
pore size 0.22 μm, PVDF, hydrophobic, filter diam. 293 mm
Expand
View Pricing
HVHP29325
Durapore
®
Screen Filter,
pore size 0.45 μm, PVDF (Hydrophilic), hydrophobic, filter diam. 293 mm
Expand
View Pricing