![D-tert-Leucine 98%](/deepweb/assets/sigmaaldrich/product/structures/428/486/8f382a47-27a3-4468-b643-aa18078cae98/640/8f382a47-27a3-4468-b643-aa18078cae98.png)
D-tert-Leucine
![D-tert-Leucine 98%](/deepweb/assets/sigmaaldrich/product/structures/428/486/8f382a47-27a3-4468-b643-aa18078cae98/640/8f382a47-27a3-4468-b643-aa18078cae98.png)
Synonym(s):
(R)-2-Amino-3,3-dimethylbutyric acid, D-α-tert-Butylglycine
Linear Formula:
(CH3)3CCH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing