Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
D-Pantothenic acid hemicalcium salt
Synonym(s):
(
R
)-(+)-
N
-(2,4-Dihydroxy-3,3-dimethyl-1-oxobutyl)-β-alanine hemicalcium salt, Calcium D-pantothenate, Vitamin B
5
Linear Formula:
HOCH
2
C(CH
3
)
2
CH(OH)CONHCH
2
CH
2
CO
2
·1/2Ca
CAS Number:
137-08-6
Molecular Weight:
238.27
EC Number:
205-278-9
Products
Product Category
vitamins (2)
Brand
Sigma-Aldrich (1)
Biological Source
Color
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Optical Isomer
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
21210
D-Pantothenic acid hemicalcium salt,
≥98.0%
Expand
View Pricing
P5155
D-Pantothenic acid hemicalcium salt,
BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Expand
View Pricing