Doxercalciferol
Synonym(s):
(5Z,7E,22E)-9,10-Secoergosta-5,7,10(19),22-tetraene-1α,3β-diol, (5Z,7E,22E)-9,10-Secoergosta-5,7,10(19),22-tetraene-1,3-diol, 1α-Hydroxyergocalciferol, 1α-Hydroxyvitamin D2, 1α-OHD2, 1-α-Hydroxyvitamin D2, 1-Hydroxyergocalciferol, TSA 840
Empirical Formula (Hill Notation):
C28H44O2
CAS Number:
Molecular Weight:
412.65
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing