Skip to Content
Merck
Docetaxel purum, ≥97.0% (HPLC)

Docetaxel

Docetaxel purum, ≥97.0% (HPLC)
Empirical Formula (Hill Notation):
C43H53NO14
CAS Number:
Molecular Weight:
807.88

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing