Skip to Content
Merck
DMT-dC(bz) Pharmadite®

DMT-dC(bz) Pharmadite®

DMT-dC(bz) Pharmadite®
Synonym(s):
DMT-dC(bz) Phosphoramidite, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxycytidine, 3′-[2-cyanoethyl N,N-?bis(1-methylethyl)phosphoramidite]
Empirical Formula (Hill Notation):
C46H52N5O8P
Molecular Weight:
833.91

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing