
(±)-Taxifolin hydrate

Synonym(s):
3,3′,4′,5,7-Pentahydroxyflavanone hydrate, Dihydroquercetin hydrate
Empirical Formula (Hill Notation):
C15H12O7 · xH2O
CAS Number:
Molecular Weight:
304.25 (anhydrous basis)
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Application