![(+)-Longifolene phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/553/627/0d21a3ce-e705-44b2-ae50-841ea893ab31/640/0d21a3ce-e705-44b2-ae50-841ea893ab31.png)
(+)-Longifolene
![(+)-Longifolene phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/553/627/0d21a3ce-e705-44b2-ae50-841ea893ab31/640/0d21a3ce-e705-44b2-ae50-841ea893ab31.png)
Synonym(s):
(+)-Longifolene, (1R,2S,7S,9S)-3,3,7-Trimethyl-8-methylenetricyclo[5.4.0.02.9]undecane, Junipene, Kuromatsuene, [1S-(1α,3aβ,4α,8aβ)]-Decahydro-4,8,8-trimethyl-9-methylene-1,4-methanoazulene
Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing