DL-Lactic acid
Synonym(s):
2-Hydroxypropionic acid
Linear Formula:
CH3CH(OH)COOH
CAS Number:
Molecular Weight:
90.08
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing