Skip to Content
Merck
Diphenhydramine-D3 solution 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Diphenhydramine-D3 solution

Diphenhydramine-D3 solution 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C17D3H18NO
CAS Number:
Molecular Weight:
258.37
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique