
Dioctyl adipate

Synonym(s):
Adipic acid dioctyl ester
Empirical Formula (Hill Notation):
C22H42O4
CAS Number:
Molecular Weight:
370.57
Products
Brand
Formula Weight
Agency
Quality Segment
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing