![Diisobutylaluminum hydride solution 1.0 M in heptane](/deepweb/assets/sigmaaldrich/product/structures/139/606/6c8d8392-05c8-400c-a391-631a1a171716/640/6c8d8392-05c8-400c-a391-631a1a171716.png)
Diisobutylaluminum hydride solution
![Diisobutylaluminum hydride solution 1.0 M in heptane](/deepweb/assets/sigmaaldrich/product/structures/139/606/6c8d8392-05c8-400c-a391-631a1a171716/640/6c8d8392-05c8-400c-a391-631a1a171716.png)
Synonym(s):
DIBAL, DIBAL-H
Linear Formula:
[(CH3)2CHCH2]2AlH
CAS Number:
Molecular Weight:
142.22
Products
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Reagent Type
Showing 1-7 of 7
Compare
Product Number
Description
Pricing