Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dicyclomine hydrochloride
Synonym(s):
2-(Diethylamino)ethyl 1-cyclohexylcyclohexane-1-carboxylate hydrochloride, [1,1-Bicyclohexyl]-1-carboxylic acid 2-(diethylamino)ethyl ester hydrochloride
Empirical Formula (Hill Notation):
C
19
H
35
NO
2
· HCl
CAS Number:
67-92-5
Molecular Weight:
345.95
EC Number:
200-671-1
Products
Product Category
bioactive small molecules (1)
Brand
Sigma-Aldrich (1)
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Special Grade
pharmaceutical primary standard (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1191000
Dicyclomine hydrochloride,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
D7909
Dicyclomine hydrochloride,
≥99% (TLC), powder
Expand
View Pricing