Dicloxacillin sodium salt monohydrate
Empirical Formula (Hill Notation):
C19H16Cl2N3NaO5S·H2O
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Activity Spectrum
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing