
Diclazuril

Synonym(s):
2,6-Dichloro-α-(4-chlorophenyl)-4-[4,5-dihydro-3,5-dioxo-1,2,4-triazin-2(3H)-yl]benzeneacetonitrile
Empirical Formula (Hill Notation):
C17H9Cl3N4O2
CAS Number:
Molecular Weight:
407.64
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing