Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dibenzyl
N
,
N
-diethylphosphoramidite
Synonym(s):
TTC
Linear Formula:
(C
2
H
5
)
2
NP(OCH
2
C
6
H
5
)
2
CAS Number:
67746-43-4
Molecular Weight:
317.36
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Technical (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
362883
Dibenzyl
N
,
N
-diethylphosphoramidite,
technical grade, 85%
Expand
View Pricing