Skip to Content
Merck
Diallyl phthalate analytical standard

Diallyl phthalate

Diallyl phthalate analytical standard
Linear Formula:
C6H4-1,2-(CO2CH2CH=CH2)2
CAS Number:
Molecular Weight:
246.26
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing