![D-Glucose 6-phosphate potassium salt ≥95% anhydrous basis (enzymatic)](/deepweb/assets/sigmaaldrich/product/structures/405/301/edb31742-bd1c-499b-90da-705c175c7b07/640/edb31742-bd1c-499b-90da-705c175c7b07.png)
D-Glucose 6-phosphate potassium salt
![D-Glucose 6-phosphate potassium salt ≥95% anhydrous basis (enzymatic)](/deepweb/assets/sigmaaldrich/product/structures/405/301/edb31742-bd1c-499b-90da-705c175c7b07/640/edb31742-bd1c-499b-90da-705c175c7b07.png)
Synonym(s):
D(+)-Glucopyranose 6-phosphate potassium salt, Robison ester
Linear Formula:
C6H12O9PK
CAS Number:
Molecular Weight:
298.23
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing