![D-Glucose-1-d1 98 atom % D](/deepweb/assets/sigmaaldrich/product/structures/797/021/bfe407d3-47ab-4e53-badc-729ae88480f4/640/bfe407d3-47ab-4e53-badc-729ae88480f4.png)
D-Glucose-1-d1
![D-Glucose-1-d1 98 atom % D](/deepweb/assets/sigmaaldrich/product/structures/797/021/bfe407d3-47ab-4e53-badc-729ae88480f4/640/bfe407d3-47ab-4e53-badc-729ae88480f4.png)
Synonym(s):
Dextrose-1-d1
Linear Formula:
HOCH2[CHOH]4CDO
CAS Number:
Molecular Weight:
181.16
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing