Skip to Content
Merck
Dextrose monohydrate 97.5-102.0% anhydrous basis, meets USP testing specifications

Dextrose monohydrate

Dextrose monohydrate 97.5-102.0% anhydrous basis, meets USP testing specifications
Synonym(s):
D-(+)-Glucose monohydrate, Dextrose monohydrate
Empirical Formula (Hill Notation):
C6H12O6 · H2O
CAS Number:
Molecular Weight:
198.17
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Physical Form
Agency
Quality Segment
Optical Isomer
Showing 1-1 of 1