Dexchlorpheniramine Maleate
Synonym(s):
S-(+)-Chlorpheniramine maleate salt, (S)-γ-(4-Chlorophenyl)-N,N-dimethyl-2-pyridinepropanamine maleate salt
Empirical Formula (Hill Notation):
C16H19ClN2 · C4H4O4
CAS Number:
Molecular Weight:
390.86
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3